Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026534.2 | NP_080810.2 | 6 | 627 | |
GenBank mRNA | AB120715.1 | - | 6 | 620 | |
AK012822.1 | - | 6 | 626 | ||
AK013985.1 | - | 6 | 622 | ||
AK028268.1 | - | 6 | 630 | ||
AK032332.1 | - | 6 | 627 | ||
AK049018.1 | - | 6 | 635 | ||
BC076632.1 | - | 6 | 627 | 30637967 |