Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033874.2 | NP_001029046.2 | 7, 8 | 591 | |
GenBank mRNA | AK004439.1 | - | 7, 8 | 589 | |
BC100481.1 | - | 7, 8 | 588 | 30917849 | |
BC109352.2 | - | 7, 8 | 578 | 40057006 |