Gene Transcripts

Gene Symbol
Zg16
Entrez Gene ID
69036
Gene Name
zymogen granule protein 16
Gene Aliases
1810010M01Rik, AI593689, ZG16p
Chromosome Location
Chr. 7: 127050156 - 127052701 on Build GRCm38
UniGene ID
Mm.21835
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026918.2 NP_081194.1 1 41
GenBank mRNA BC031800.1 - 1 41

View Full Product Details