Gene Transcripts

Gene Symbol
Rasgef1c
Entrez Gene ID
74563
Gene Name
RasGEF domain family, member 1C
Gene Aliases
9130006A14Rik
Chromosome Location
Chr. 11: 49901816 - 49980223 on Build GRCm38
UniGene ID
Mm.258798
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029004.1 NP_083280.1 9, 10 1117
GenBank mRNA AK018593.1 - 9, 10 1106
AK039330.1 - 1481
AK047088.1 - 8, 9 996

View Full Product Details