Gene Transcripts
- Gene Symbol
- Sacm1l
- Entrez Gene ID
- 83493
- Gene Name
- SAC1 (suppressor of actin mutations 1, homolog)-like (S. cerevisiae)
- Gene Aliases
- SAC1, Sac1p, mKIAA0851
- Chromosome Location
- Chr. 9: 123529882 - 123592598 on Build GRCm38
- UniGene ID
- Mm.273671
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030692.4 |
NP_109617.1
|
9, 10 |
780 |
|
GenBank mRNA |
AJ245720.1 |
- |
9, 10 |
910 |
|
AJ297526.1 |
- |
9, 10 |
781 |
|
AK089174.1 |
- |
9, 10 |
864 |
|
AK122390.1 |
- |
9, 10 |
845 |
|
AK154759.1 |
- |
9, 10 |
880 |
|
AK161111.1 |
- |
9, 10 |
814 |
|
AK171080.1 |
- |
9, 10 |
864 |
|
BC117752.1 |
- |
9, 10 |
781 |
40090556 |
BC117753.1 |
- |
9, 10 |
781 |
40090558 |
View Full Product Details