Gene Transcripts

Gene Symbol
Pcdhgb8
Entrez Gene ID
93705
Gene Name
protocadherin gamma subfamily B, 8
Gene Aliases
Chromosome Location
Chr. 18: 37761801 - 37841873 on Build GRCm38
UniGene ID
Mm.247203
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033580.2 NP_291058.1 1 715
GenBank mRNA AY013810.1 - 1 637
BC139099.1 - 1 715 8862121
BC139100.1 - 1 715 8862122

View Full Product Details