Gene Transcripts

Gene Symbol
Ugt2b34
Entrez Gene ID
100727
Gene Name
UDP glucuronosyltransferase 2 family, polypeptide B34
Gene Aliases
AI788959
Chromosome Location
Chr. 5: 86889770 - 86906937 on Build GRCm38
UniGene ID
Mm.281844
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153598.2 NP_705826.1 6 1499
GenBank mRNA AK145951.1 - 5 2770
BC028826.1 - 6 1499 5098890

View Full Product Details