Gene Transcripts

Gene Symbol
Slc34a3
Entrez Gene ID
142681
Gene Name
solute carrier family 34 (sodium phosphate), member 3
Gene Aliases
AI649385, Npt2c, Npt2c-v1, NptIIc, naPi-2c
Chromosome Location
Chr. 2: 25228897 - 25234271 on Build GRCm38
UniGene ID
Mm.346652
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080854.3 NP_543130.2 7 703
GenBank mRNA AB054999.1 - 7 683
BC125326.1 - 6 636 40129841
BC131997.1 - 6 636 40130274

View Full Product Details