Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164171.1 | NP_001157643.1 | 9 | 961 | |
NM_010856.4 | NP_034986.1 | 9 | 897 | ||
GenBank mRNA | AK142232.1 | - | 9 | 934 | |
BC061145.1 | - | 9 | 930 | ||
BC090979.1 | - | 9 | 930 | ||
BC110700.1 | - | 9 | 943 | 6786621 | |
M76599.1 | - | 7 | 759 | ||
M76600.1 | - | 7 | 759 | ||
M76601.1 | - | 9 | 861 |