Gene Transcripts

Gene Symbol
Vmn2r89
Entrez Gene ID
22301
Gene Name
vomeronasal 2, receptor 89
Gene Aliases
V2r10, V2r11
Chromosome Location
Chr. 14: 51451962 - 51461293 on Build GRCm38
UniGene ID
Mm.482572
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009487.1 NP_033513.1 4 1406
GenBank mRNA AF011421.1 - 4 1406
BC145855.1 - 4 1367 40131158

View Full Product Details