Gene Transcripts
- Gene Symbol
- Txlng
- Entrez Gene ID
- 353170
- Gene Name
- taxilin gamma
- Gene Aliases
- 4932441K18Rik, Fiat, Lrp
- Chromosome Location
- Chr. X: 162778917 - 162829454 on Build GRCm38
- UniGene ID
- Mm.270186
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290776.1 |
NP_001277705.1
|
10, 11 |
1204 |
|
NM_178935.5 |
NP_849266.1
|
9, 10 |
1351 |
|
GenBank mRNA |
AK030100.1 |
- |
9, 10 |
1298 |
|
AK031023.1 |
- |
7, 8 |
2890 |
|
AK031783.1 |
- |
9, 10 |
1265 |
|
AK044130.1 |
- |
9, 10 |
1300 |
|
BC117898.1 |
- |
10, 11 |
1145 |
40096960 |
BC117899.1 |
- |
10, 11 |
1145 |
40096967 |
DQ316984.1 |
- |
9, 10 |
1351 |
|
DQ320011.1 |
- |
10, 11 |
1204 |
|
View Full Product Details