Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166350.1 | NP_001159822.1 | 3 | 807 | |
NM_199314.2 | NP_955018.2 | 3 | 821 | ||
GenBank mRNA | AK149420.1 | - | 3 | 785 | |
BC024087.1 | - | 3 | 799 | 5101271 | |
BC096459.1 | - | 4 | 713 | ||
BC096585.1 | - | 3 | 768 | 5097495 |