Gene Transcripts

Gene Symbol
Krtap16-1
Entrez Gene ID
100504183
Gene Name
keratin associated protein 16-1
Gene Aliases
Gm11570, OTTMUSG00000002209
Chromosome Location
Chr. 11: 99984703 - 99986593 on Build GRCm38
UniGene ID
Mm.426560
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256057.1 NP_001242986.1 1 1335

View Full Product Details