Gene Transcripts

Gene Symbol
Ctsr
Entrez Gene ID
56835
Gene Name
cathepsin R
Gene Aliases
AI428350
Chromosome Location
Chr. 13: 61159215 - 61164188 on Build GRCm38
UniGene ID
Mm.315715
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020284.1 NP_064680.1 4 425
GenBank mRNA AF245399.1 - 4 425
AK005429.1 - 4 445
AK014432.1 - 4 441
AY014778.1 - 4 384
BC100338.1 - 4 420 30712889

View Full Product Details