Gene Transcripts

Gene Symbol
Pdgfrl
Entrez Gene ID
68797
Gene Name
platelet-derived growth factor receptor-like
Gene Aliases
1110039P19Rik, AV013190
Chromosome Location
Chr. 8: 40926233 - 40990782 on Build GRCm38
UniGene ID
Mm.284246
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026840.3 NP_081116.3 2 282
GenBank mRNA AK004179.1 - 2 174
AK081847.1 - 2 308
AK132625.1 - 2 283
BC058275.1 - 2 274 6515674

View Full Product Details