Gene Transcripts

Gene Symbol
Znrd1as
Entrez Gene ID
76416
Gene Name
Znrd1 antisense
Gene Aliases
1700022C21Rik, AI746295, Znrd1-as
Chromosome Location
Chr. 17: 36958592 - 36965625 on Build GRCm38
UniGene ID
Mm.26684
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029602.1 NP_083878.1 3 749
GenBank mRNA AK006236.1 - 3 742
AK161503.1 - 3 743
BC027007.1 - 3 749 5365474

View Full Product Details