Gene Transcripts

Gene Symbol
Klk1b4
Entrez Gene ID
18048
Gene Name
kallikrein 1-related pepidase b4
Gene Aliases
Ngfa, mGk-4
Chromosome Location
Chr. 7: 44207435 - 44211754 on Build GRCm38
UniGene ID
Mm.440886
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010915.3 NP_035045.2 729
GenBank mRNA BC034518.1 - 729
M11434.1 - 725

View Full Product Details