Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001025571.2 | NP_001020742.1 | 3 | 669 | |
NM_021309.3 | NP_067284.1 | 3 | 669 | ||
GenBank mRNA | AF203343.1 | - | 3 | 676 | |
AK037489.1 | - | 3 | 391 | ||
AK172182.1 | - | 3 | 421 | ||
BC034847.1 | - | 3 | 401 | 3327113 | |
U69490.1 | - | 3 | 379 |