Gene Transcripts

Gene Symbol
Akna
Entrez Gene ID
100182
Gene Name
AT-hook transcription factor
Gene Aliases
AI597013
Chromosome Location
Chr. 4: 63367127 - 63410758 on Build GRCm38
UniGene ID
Mm.387725
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK089194.1 - 735
AK129468.1 - 727
AK158313.1 - 3 639
AK159547.1 - 810
AK170885.1 - 702
BC092227.1 - 2 591 30531788

View Full Product Details