Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133685.2 | NP_598446.2 | 2 | 300 | |
GenBank mRNA | AB232617.1 | - | 2 | 92 | |
AK051270.1 | - | 2 | 220 | ||
AK156677.1 | - | 2 | 301 | ||
AK159095.1 | - | 2 | 189 | ||
AK166151.1 | - | 2 | 191 | ||
BC013063.1 | - | 2 | 229 | 3497938 |