Gene Transcripts

Gene Symbol
Me3
Entrez Gene ID
109264
Gene Name
malic enzyme 3, NADP(+)-dependent, mitochondrial
Gene Aliases
1700020C08Rik, B230207H15Rik
Chromosome Location
Chr. 7: 89632197 - 89854359 on Build GRCm38
UniGene ID
Mm.334011
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181407.4 NP_852072.2 6, 7 900
GenBank mRNA AK032196.1 - 6, 7 901
AK080798.1 - 6, 7 833
BC099935.1 - 6, 7 996 30097608

View Full Product Details