Gene Transcripts
- Gene Symbol
- Tpbpb
- Entrez Gene ID
- 116913
- Gene Name
- trophoblast specific protein beta
- Gene Aliases
- 1600012N09Rik
- Chromosome Location
- Chr. 13: 60901295 - 60904847 on Build GRCm38
- UniGene ID
- Mm.443229
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026429.4 |
NP_080705.1
|
2, 3 |
187 |
|
GenBank mRNA |
AK005433.1 |
- |
2, 3 |
187 |
|
AK018793.1 |
- |
2, 3 |
186 |
|
AK018794.1 |
- |
2, 3 |
185 |
|
AK018795.1 |
- |
2, 3 |
187 |
|
AK018798.1 |
- |
2, 3 |
187 |
|
AK018799.1 |
- |
2, 3 |
187 |
|
AK018804.1 |
- |
2, 3 |
188 |
|
AK018805.1 |
- |
2, 3 |
187 |
|
AK018808.1 |
- |
2, 3 |
188 |
|
AK018809.1 |
- |
2, 3 |
186 |
|
AK018810.1 |
- |
2, 3 |
187 |
|
AK018820.1 |
- |
2, 3 |
187 |
|
AK018824.1 |
- |
2, 3 |
187 |
|
AK018827.1 |
- |
2, 3 |
186 |
|
AY034576.1 |
- |
2, 3 |
162 |
|
BC099441.1 |
- |
2, 3 |
172 |
|
BC100333.1 |
- |
2, 3 |
172 |
30787126 |
View Full Product Details