Gene Transcripts
- Gene Symbol
- Drosha
- Entrez Gene ID
- 14000
- Gene Name
- drosha, ribonuclease type III
- Gene Aliases
- 1110013A17Rik, AI874853, Etohi2, Rn3, Rnasen
- Chromosome Location
- Chr. 15: 12824804 - 12935291 on Build GRCm38
- UniGene ID
- Mm.293142
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130149.1 |
NP_001123621.1
|
24, 25 |
3325 |
|
NM_026799.3 |
NP_081075.3
|
24, 25 |
3321 |
|
GenBank mRNA |
AF440766.1 |
- |
7, 8 |
666 |
|
AF533013.1 |
- |
20, 21 |
1989 |
|
AK144147.1 |
- |
24, 25 |
3280 |
|
AK148640.1 |
- |
24, 25 |
3244 |
|
BC040801.1 |
- |
9, 10 |
944 |
|
BC050057.1 |
- |
10, 11 |
1043 |
|
BC055696.1 |
- |
13, 14 |
1266 |
|
BC057687.1 |
- |
17, 18 |
1753 |
|
BC060265.1 |
- |
21, 22 |
2620 |
|
BC088999.1 |
- |
24, 25 |
3259 |
6417566 |
View Full Product Details