Gene Transcripts

Gene Symbol
Hoxc8
Entrez Gene ID
15426
Gene Name
homeobox C8
Gene Aliases
D130011F21Rik, Hox-3.1
Chromosome Location
Chr. 15: 102990539 - 102994254 on Build GRCm38
UniGene ID
Mm.6167
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010466.2 NP_034596.1 1 346
GenBank mRNA AB221617.1 - 1 109
X07439.1 - 1 346
X07646.1 - 1 574

View Full Product Details