Gene Transcripts

Gene Symbol
Rrp1
Entrez Gene ID
18114
Gene Name
ribosomal RNA processing 1 homolog (S. cerevisiae)
Gene Aliases
AL033305, AL033306, AU018604, NNP-1, Nnp1
Chromosome Location
Chr. 10: 78400362 - 78413043 on Build GRCm38
UniGene ID
Mm.38344
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010925.2 NP_035055.2 7 658
GenBank mRNA AF312394.1 - 6 421
AK077540.1 - 7 583
BC022910.1 - 6 524 4165783
BC065993.1 - 7 620 5684672
BC065994.1 - 7 578 5685269
U79773.1 - 7 610
U79774.1 - 7 610

View Full Product Details