Gene Transcripts
- Gene Symbol
- Plp1
- Entrez Gene ID
- 18823
- Gene Name
- proteolipid protein (myelin) 1
- Gene Aliases
- DM20, Plp, jimpy, jp, msd, rsh
- Chromosome Location
- Chr. X: 136822746 - 136838582 on Build GRCm38
- UniGene ID
- Mm.1268
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290561.1 |
NP_001277490.1
|
6, 7 |
815 |
|
NM_001290562.1 |
NP_001277491.1
|
5, 6 |
846 |
|
NM_011123.3 |
NP_035253.1
|
6, 7 |
920 |
|
GenBank mRNA |
AK077564.1 |
- |
6, 7 |
774 |
|
AK134554.1 |
- |
6, 7 |
911 |
|
AK160872.1 |
- |
6, 7 |
915 |
|
BC027010.1 |
- |
6, 7 |
830 |
5364736 |
M14674.1 |
- |
5, 6 |
754 |
|
M15442.1 |
- |
6, 7 |
899 |
|
M16472.1 |
- |
5, 6 |
649 |
|
M20752.1 |
- |
4, 5 |
680 |
|
View Full Product Details