Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011133.2 | NP_035263.1 | 9, 10 | 716 | |
GenBank mRNA | AF036898.1 | - | 9, 10 | 689 | |
AK077592.1 | - | 9, 10 | 717 | ||
AK172024.1 | - | 9, 10 | 711 | ||
BC052170.1 | - | 9, 10 | 705 | ||
BC063772.1 | - | 9, 10 | 705 | 30147626 |