Gene Transcripts

Gene Symbol
Scrg1
Entrez Gene ID
20284
Gene Name
scrapie responsive gene 1
Gene Aliases
AW124307
Chromosome Location
Chr. 8: 57455923 - 57477585 on Build GRCm38
UniGene ID
Mm.12886
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009136.3 NP_033162.1 2 313
GenBank mRNA AJ223206.1 - 2 313
AK046213.1 - 2 182
BC058532.1 - 201
CT010374.1 - 1 97

View Full Product Details