Gene Transcripts

Gene Symbol
Ppp2r3a
Entrez Gene ID
235542
Gene Name
protein phosphatase 2, regulatory subunit B'', alpha
Gene Aliases
3222402P14Rik, A730042E07
Chromosome Location
Chr. 9: 101104989 - 101251832 on Build GRCm38
UniGene ID
Mm.335386
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161362.3 NP_001154834.1 3 2774
NM_172144.3 NP_742156.2 2 614
GenBank mRNA AK035077.1 - 2 394
AK076163.1 - 2 1826
AK132175.1 - 2 615
AK169510.1 - 2 547
BC042841.1 - 2 1736
BC042842.1 - 2 2209
BC151170.1 - 2 2601 9088059
BC151171.1 - 2 2596 9088060

View Full Product Details