Gene Transcripts

Gene Symbol
Thada
Entrez Gene ID
240174
Gene Name
thyroid adenoma associated
Gene Aliases
6530405K19, A930021O22, AU019899, BC052885, GITA
Chromosome Location
Chr. 17: 84190055 - 84466261 on Build GRCm38
UniGene ID
Mm.246767
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183021.3 NP_898842.2 29 4306
GenBank mRNA AK173258.1 - 14 1816
BC052885.1 - 4 446
EF222207.1 - 29 4214

View Full Product Details