Gene Transcripts

Gene Symbol
Lzts3
Entrez Gene ID
241638
Gene Name
leucine zipper, putative tumor suppressor family member 3
Gene Aliases
Prosapip1
Chromosome Location
Chr. 2: 130632765 - 130642844 on Build GRCm38
UniGene ID
Mm.440130
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291027.1 NP_001277956.1 3 575
NM_197945.4 NP_922936.3 3 575
GenBank mRNA AK148251.1 - 3 534
AK161711.1 - 3 529
BC049892.1 - 1 136

View Full Product Details