Gene Transcripts
- Gene Symbol
- Rsrp1
- Entrez Gene ID
- 27981
- Gene Name
- arginine/serine rich protein 1
- Gene Aliases
- 2700043I21Rik, D4Ucla2, D4Wsu53e
- Chromosome Location
- Chr. 4: 134923625 - 134927370 on Build GRCm38
- UniGene ID
- Mm.331964
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023665.3 |
NP_076154.3
|
2, 3 |
759 |
|
GenBank mRNA |
AB041555.1 |
- |
2, 3 |
739 |
|
AK003799.1 |
- |
2, 3 |
760 |
|
AK012363.1 |
- |
2, 3 |
760 |
|
AK028145.1 |
- |
2, 3 |
761 |
|
AK029529.1 |
- |
2, 3 |
759 |
|
AK150024.1 |
- |
2, 3 |
760 |
|
AK150665.1 |
- |
2, 3 |
739 |
|
AK152736.1 |
- |
2, 3 |
739 |
|
AK160993.1 |
- |
2, 3 |
760 |
|
AK169598.1 |
- |
2, 3 |
761 |
|
AK169760.1 |
- |
2, 3 |
760 |
|
AK178549.1 |
- |
1, 2 |
105 |
|
AK206224.1 |
- |
1, 2 |
105 |
|
BC043057.1 |
- |
2, 3 |
744 |
6419597 |
BC046231.1 |
- |
2, 3 |
661 |
6813388 |
BC054728.1 |
- |
2, 3 |
732 |
6419723 |
BC056986.1 |
- |
2, 3 |
693 |
6816649 |
View Full Product Details