Gene Transcripts

Gene Symbol
Samd12
Entrez Gene ID
320679
Gene Name
sterile alpha motif domain containing 12
Gene Aliases
A830094I09Rik
Chromosome Location
Chr. 15: 53399119 - 53902536 on Build GRCm38
UniGene ID
Mm.442455
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177225.4 NP_796199.1 2 123
GenBank mRNA AK032588.1 - 2 91
AK044142.1 - 2 124
AK080536.1 - 2 91
BC119383.1 - 2 124 8734132
BC119385.1 - 2 124 8734134
BC144982.1 - 2 124 9053530

View Full Product Details