Gene Transcripts

Gene Symbol
Obp2b
Entrez Gene ID
383678
Gene Name
odorant binding protein 2B
Gene Aliases
Lcn14, OTTMUSG00000012631
Chromosome Location
Chr. 2: 25714105 - 25740115 on Build GRCm38
UniGene ID
Mm.464561
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099301.1 NP_001092771.1 2 219

View Full Product Details