Gene Transcripts

Gene Symbol
Prr7
Entrez Gene ID
432763
Gene Name
proline rich 7 (synaptic)
Gene Aliases
XM_484263
Chromosome Location
Chr. 13: 55464267 - 55473155 on Build GRCm38
UniGene ID
Mm.315457
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030296.4 NP_001025467.1 3 1180
GenBank mRNA AK133129.1 - 3 1140
BC138854.1 - 2 848 8861876
BC138856.1 - 2 848 8861878

View Full Product Details