Gene Transcripts
- Gene Symbol
- Wsb2
- Entrez Gene ID
- 59043
- Gene Name
- WD repeat and SOCS box-containing 2
- Gene Aliases
- AA673511, Swip2, WSB-2
- Chromosome Location
- Chr. 5: 117357305 - 117378589 on Build GRCm38
- UniGene ID
- Mm.28489
- Species
- Mus musculus
- Gene Symbol
- Gm10428
- Entrez Gene ID
- 100049172
- Gene Name
- predicted gene 10428
- Gene Aliases
- ENSMUSG00000072843
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.379767
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK089738.1 |
- |
1 |
366 |
|
View Full Product Details