Gene Transcripts

Gene Symbol
Ttll2
Entrez Gene ID
100216474
Gene Name
tubulin tyrosine ligase-like family, member 2
Gene Aliases
EG625850, ENSMUSG00000079722
Chromosome Location
Chr. 17: 7350625 - 7364035 on Build GRCm38
UniGene ID
Mm.477432
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098267.1 NP_001091737.1 2 204
GenBank mRNA AM690745.1 - 2 204

View Full Product Details