Gene Transcripts

Gene Symbol
Vmn1r77
Entrez Gene ID
628946
Gene Name
vomeronasal 1 receptor 77
Gene Aliases
Gm6935
Chromosome Location
Chr. 7: 12036846 - 12042219 on Build GRCm38
UniGene ID
Mm.484974
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166731.1 NP_001160203.1 1 764

View Full Product Details