Gene Transcripts

Gene Symbol
Marc1
Entrez Gene ID
66112
Gene Name
mitochondrial amidoxime reducing component 1
Gene Aliases
1300013F15Rik, Mosc1
Chromosome Location
Chr. 1: 184786767 - 184811308 on Build GRCm38
UniGene ID
Mm.272457
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290273.1 NP_001277202.1 5 977
GenBank mRNA AK004989.1 - 5 874
AK011722.1 - 5 858
BC028441.1 - 3 326

View Full Product Details