Gene Transcripts

Gene Symbol
Ubl5
Entrez Gene ID
66177
Gene Name
ubiquitin-like 5
Gene Aliases
1110030M22Rik, beacon
Chromosome Location
Chr. 9: 20643318 - 20646790 on Build GRCm38
UniGene ID
Mm.390165
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025401.3 NP_079677.1 4, 5 1756
GenBank mRNA AK002730.1 - 4, 5 265
AK003992.1 - 4, 5 272
AK007726.1 - 4, 5 237
AK009854.1 - 4, 5 272
AK012803.1 - 4, 5 254
AK051149.1 - 4, 5 1757
AK154036.1 - 3, 4 2348
BC028498.1 - 4, 5 226 1400865

View Full Product Details