Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290182.1 | NP_001277111.1 | 1, 2 | 125 | |
NM_024437.4 | NP_077757.2 | 1, 2 | 125 | ||
NM_024446.5 | NP_077766.3 | 1, 2 | 125 | ||
GenBank mRNA | AF338424.1 | - | 1, 2 | 85 | |
AK008824.1 | - | 1, 2 | 52 | ||
AK011172.1 | - | 1, 2 | 59 | ||
BC033046.1 | - | 1, 2 | 57 | ||
BC069843.1 | - | 1, 2 | 81 | 5102473 | |
CA460240.1 | - | 1, 2 | 157 |