Gene Transcripts

Gene Symbol
Mak16
Entrez Gene ID
67920
Gene Name
MAK16 homolog (S. cerevisiae)
Gene Aliases
2600016B03Rik, AI314911, Rbm13
Chromosome Location
Chr. 8: 31159468 - 31168724 on Build GRCm38
UniGene ID
Mm.289818
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026453.3 NP_080729.1 5 405
GenBank mRNA AK015369.1 - 5 446
AK075982.1 - 5 406
AK075983.1 - 5 406
BC013079.1 - 5 420 3499754

View Full Product Details