Gene Transcripts

Gene Symbol
Nat8
Entrez Gene ID
68396
Gene Name
N-acetyltransferase 8 (GCN5-related, putative)
Gene Aliases
0610037O16Rik, CCNAT, Cml4
Chromosome Location
Chr. 6: 85830387 - 85836636 on Build GRCm38
UniGene ID
Mm.154782
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023455.3 NP_075944.1 2 791
GenBank mRNA AF163317.1 - 2 748
AK002784.1 - 2 732
BC019517.1 - 2 983 4236543

View Full Product Details