Gene Transcripts

Gene Symbol
Yipf7
Entrez Gene ID
75581
Gene Name
Yip1 domain family, member 7
Gene Aliases
2310016N21Rik, Yip1b
Chromosome Location
Chr. 5: 69516670 - 69542647 on Build GRCm38
UniGene ID
Mm.154776
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023784.5 NP_076273.1 3 294
GenBank mRNA AF217188.1 - 2 206
AK009405.1 - 3 245
BC089576.1 - 3 279 6758950

View Full Product Details