Gene Transcripts

Gene Symbol
Asb8
Entrez Gene ID
78541
Gene Name
ankyrin repeat and SOCS box-containing 8
Gene Aliases
4930539L19Rik, AI788835, C430011H06Rik
Chromosome Location
Chr. 15: 98134637 - 98165602 on Build GRCm38
UniGene ID
Mm.200766
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170710.1 NP_001164181.1 4 617
NM_001170711.1 NP_001164182.1 4 623
NM_030121.4 NP_084397.3 4 613
GenBank mRNA AF398969.1 - 4 540
AK016001.1 - 4 671
AK030168.1 - 4 556
AK087299.1 - 4 575
AK218426.1 - 1 20
BC025106.1 - 4 537 4194829

View Full Product Details