Gene Transcripts

Gene Symbol
Bcl9l
Entrez Gene ID
80288
Gene Name
B cell CLL/lymphoma 9-like
Gene Aliases
B9L, BC003321, BCL9-2, DLNB11
Chromosome Location
Chr. 9: 44483113 - 44511901 on Build GRCm38
UniGene ID
Mm.370270
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030256.2 NP_084532.2 7 2952
GenBank mRNA AB128033.1 - 6 2738
AY296058.1 - 7 2952
BC082304.1 - 6 3086 6856460
BC119466.2 - 2 560
BC119467.2 - 2 552

View Full Product Details