Gene Transcripts

Gene Symbol
Alg3
Entrez Gene ID
287983
Gene Name
ALG3, alpha-1,3- mannosyltransferase
Gene Aliases
Chromosome Location
Chr. 11: 83986212 - 83991708 on Build Rnor_6.0
UniGene ID
Rn.106814
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142363.1 NP_001135835.1 6 810
XM_006248558.2 XP_006248620.1 6 828
GenBank mRNA BC088475.1 - 5 693
FM075565.1 - 5 622

View Full Product Details