Gene Transcripts

Gene Symbol
Adh6a
Entrez Gene ID
295498
Gene Name
alcohol dehydrogenase 6A (class V)
Gene Aliases
Chromosome Location
Chr. 2: 243580381 - 243599357 on Build Rnor_6.0
UniGene ID
Rn.137460
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106475.1 NP_001099945.1 3 302
XM_006233371.2 XP_006233433.1 4 384

View Full Product Details