Gene Transcripts

Gene Symbol
Cachd1
Entrez Gene ID
298267
Gene Name
cache domain containing 1
Gene Aliases
RGD1310770
Chromosome Location
Chr. 5: 119596726 - 119832326 on Build Rnor_6.0
UniGene ID
Rn.64271
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191758.1 NP_001178687.1 12 1731

View Full Product Details