Gene Transcripts

Gene Symbol
Rasgef1b
Entrez Gene ID
100361238
Gene Name
RasGEF domain family, member 1B
Gene Aliases
RGD1562220
Chromosome Location
Chr. 14: 11994178 - 12028513 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002728083.4 XP_002728129.2 6 858
XM_008770077.1 XP_008768299.1 6 855

View Full Product Details